missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser

Product Code. 10068790
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
100 to 500 mL
2 to 10 mL
20 to 100 mL
40 to 200 mL
6 to 30 mL
60 to 300 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10068790

Brand: Poulten Graf™ 101 08033

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Poulten Graf™ Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.

For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.

  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With solid PTFE encased dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment

Specifications

Product Type Fortuna Optifix Basic Dispenser
Color Clear
Material PTFE, Polypropylene, Fiberglass, Teflon, Ceramic
Volume (Metric) 1 to 5 mL
For Use With (Application) For dispensing aqueous solutions, acids and crystallizing liquids
Quantity Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.