Learn More
Bohlender™ PTFE Propeller Stirrer Shafts
Brand: Bohlender™ C378-20
This item is not returnable.
View return policy
Specifications
10 mm | |
600 mm | |
Propeller Stirrer Shaft |
The product is sucked bottom-up, good axial flow with low shear force. | |
1 Each |