missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer are a fully molded product with a retreat angle of 30°.
Brand: Fisherbrand™
Additional Details : Weight : 0.13500kg
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer | |
8 mm | |
1 |
50 mm | |
300 x 8 mm |