missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer are a fully molded product with a retreat angle of 30°.
Brand: Fisherbrand™
Additional Details : Weight : 0.20370kg
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer | |
10 mm | |
10 mm |
50 mm | |
500 x 10 mm | |
1 |