Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand: Cole-Parmer™ 3053806
This item is not returnable.
View return policy
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
10 | |
3.81 cm | |
PTFE | |
PTFE Gasket |