Learn More
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
For grease-free sealing of ground joints and plugs, extra tight and extra clean. The alternative to ground collar, grip collar sleeve and stopper made of PTFE.
Brand:  Glindemann™ K10
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10268410
  • POA / Pack of 50
Estimated Shipment: 02-09-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
  • Solvent leak rate in containers <0.1 to 0.03mg/d.
  • Temperature resistant from -200 to +300°C.
  • The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
  • Other sizes on request.
Specifications
Specifications
PTFE
NS 10