missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ PTFE Spinbar

OHAUS™ PTFE Stirbar for stirring volumes in vessels such as vials and tubes.

Brand:  OHAUS™ 30400148

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 15702569

  • £11.20 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Constructed of PTFE
  • 10 cm spinbar includes a pivot ring
Specifications

Specifications

Spinbar 3.8 cm PTFE
White
PTFE
3.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers