Learn More
Fisherbrand™ PTFE Stir Bar with Removable Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bar with Removable Ring are autoclavable and include a pivot ring.
Brand:  Fisherbrand™ 001.17102
 View more versions of this product
Product Code. 10500482
  • £17.45 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guaranteed inert and leak-free
  • Isostatically molded in pure, FDA approved, PTFE
  • Individually polished to stay cleaner for longer
Specifications
Specifications
Autoclavable
White
102 x 16 x 16mm
16mm
16 mm
1/Ea.
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
Containers & Vessels / Stirrer Hot Plates
PTFE Ring
102 mm
Cylinder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ PTFE Stir Bar with Removable Ring > Dimensions (Dia. x L): 16 dia.x 102mmL