Learn More
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
£6.57 - £50.26
Specifications
Autoclavable | Yes |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Color | White |
Description | PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring |
Shape | Cylinder, Pivot ring |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
11898862
|
Fisherbrand™
|
60 mm | 10 mm |
£31.10
Pack of 10 |
In Stock
Log in to see stock. |
||||
11888862
|
Fisherbrand™
|
45 mm | 8 mm |
£36.69
Pack of 10 |
In Stock
Log in to see stock. |
||||
16265811
|
Fisherbrand™
|
70 mm | 10 mm |
£42.31
Pack of 10 |
In Stock
Log in to see stock. |
||||
12661156
|
Fisherbrand™
|
30 mm | 6 mm |
£18.99
Pack of 10 |
In Stock
Log in to see stock. |
||||
12651156
|
Fisherbrand™
|
12 mm | 6 mm |
£14.95
Pack of 10 |
In Stock
Log in to see stock. |
||||
11868862
|
Fisherbrand™
|
35 mm | 6 mm |
£30.03
Pack of 10 |
In Stock
Log in to see stock. |
||||
11868962
|
Fisherbrand™
|
75 mm | 13 mm |
£50.26
Pack of 5 |
In Stock
Log in to see stock. |
||||
11878862
|
Fisherbrand™
|
40 mm | 8 mm |
£27.20
Pack of 10 |
In Stock
Log in to see stock. |
||||
11808972
|
Fisherbrand™
|
25 mm | 10 mm |
£39.10
Pack of 10 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
11848862
|
Fisherbrand™
|
25 mm | 6 mm |
£22.50
Pack of 10 |
In Stock
Log in to see stock. |
||||
11838862
|
Fisherbrand™
|
20 mm | 6 mm |
£21.35
Pack of 10 |
In Stock
Log in to see stock. |
||||
11898962
|
Fisherbrand™
|
15 mm | 8 mm |
£31.11
Pack of 10 |
In Stock
Log in to see stock. |
||||
11818862
|
Fisherbrand™
|
12 mm | 4.5 mm |
£18.10
Pack of 10 |
In Stock
Log in to see stock. |
||||
11808862
|
Fisherbrand™
|
8 mm | 3 mm |
£17.35
Pack of 10 |
In Stock
Log in to see stock. |
||||
11517812
|
Fisherbrand™
|
35 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12379507
|
Fisherbrand™
|
15 mm | 4.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11888962
|
Fisherbrand™
|
13 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11527812
|
Fisherbrand™
|
40 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10373592
|
Fisherbrand™
|
70 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12641156
|
Fisherbrand™
|
50 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11507812
|
Fisherbrand™
|
25 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11597802
|
Fisherbrand™
|
20 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11527792
|
Fisherbrand™
|
30 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10018131
|
Fisherbrand™
|
60 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12359507
|
Fisherbrand™
|
12 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11587802
|
Fisherbrand™
|
12 mm | 4.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11537812
|
Fisherbrand™
|
45 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12349507
|
Fisherbrand™
|
8 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Provides stable stirring action. FisherbrandTM PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
- Stable, general-purpose magnetic stir bar
- The flat surfaces provide a greater mixing action.
- Autoclavable
Specifications
Yes | |
White | |
Cylinder, Pivot ring |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring | |
Containers & Vessels / Stirrer Hot Plates |