missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Triangle with Rib Stir Bar

Fisherbrand™ PTFE Triangle with Rib Stir Bars are effective fro dissolving solids and mixing sediments because of the scraper like action. They have excellent turbulence at all speeds and can be used on uneven surfaces.

Brand:  Fisherbrand™ 001.1812

 View more versions of this product

Product Code. 16205761

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Encapsulated PTFE
  • Alnico V Magnet Core
  • Polished finish to reduce pick-up and contamination
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Easy to clean
Specifications

Specifications

Fisherbrand™ PTFE Triangle with Rib Stir Bar
-200°C to +280°C
12 mm
1
Uneven surfaces
6 mm
PTFE with Alnico V Magnet Core
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers