missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Hose

Product Code. p-8002508 Shop All Bürkle Products
Click to view available options
Flow Rate:
270 mL/stroke
400 mL/stroke
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11901591

Brand: Buerkle™ 56060601

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides high chemical resistance and safe transfer, even with liquids which attack conventional materials such as PP or PVC.

  • Medium-contacting parts manufactured from PTFE
  • Suitable for transferring ultra-pure liquids
  • Stopcock prevents hazardous vapours from escaping and stops dripping after transfer
  • Discharge hose is 1.2m in length
  • Immersion tube with a diameter of 32mm
  • All valves are made of solid PTFE/PVDF
  • Top quality, 3 years’ warranty
TRUSTED_SUSTAINABILITY

Specifications

Description Immersion depth of 60 cm
Includes Discharge Tube
No. per Pack 1 Ea.
Material PTFE
Flow Rate 270 mL/stroke
Type Barrel Pump
Applications Liquids which attack conventional materials such as Polypropylene or PVC can now be safely transferred (except for elemental Fluorine), transferring ultra-pure liquids
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.