missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Tube
The PTFE Ultrapure Barrel Pump with Discharge Tube is suitable for the transfer of ultra-pure liquids and those that attack conventional materials such as PP or PVC. The stopcock prevents vapours from escaping, and stops dripping after transfer.
Brand: Buerkle™ 5606-0600
This item is not returnable.
View return policy
Description
- Barrel pump with rigid discharge tube with stopcock made of PTFE
- The barrel pump is also suitable for transferring ultra-pure liquids
- The stopcock prevents hazardous vapours from escaping, and stops dripping after transfer
- Immersion tube Ø 32 mm
- All valves are made of solid PTFE/PVDF
- Top quality - 3 years' warranty
Specifications
Ultra-Pure Liquid Transfer, chemically aggressive liquids | |
PTFE | |
Chemicals | |
Immersion depth of 60cm | |
Rigid discharge tube with stopcock made of PTFE |
60 cm | |
1.2 m | |
Barrel Pump | |
270 mL/stroke | |
Tube diameter of 32 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Tube > Flow Rate: 270mL/stroke; Immersion depth: 60 cm
Spot an opportunity for improvement?Share a Content Correction