missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ PTFE US Sleeves with Gripping Ring

Provides for high-vacuum tight sealing with a low leak rate.

Brand:  Bohlender™ H935-09

Product Code. 10691322

This item is not returnable. View return policy

Description

Description

  • Providing temperature resistance from −200° to +250°C
  • Universal chemical resistance
  • FDA conformity
  • Vacuum suitable
  • Reusable
  • Made of PTFE with gripping ring and sealing rings on the outside
  • Low leak rate of less than 1 x 0.0001 mbar
  • Solid construction for continued use
  • For US standard size of either 29/42 or 45/50
Specifications

Specifications

High-vacuum sealing
−200° to +250°C
Sleeve with Gripping Ring
PTFE
US 45/50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers