missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGIR (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577695
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.Specifications
PTGIR (extracellular) | |
Polyclonal | |
Unconjugated | |
PTGIR | |
Prostanoid Prostacyclin Receptor (extracellular) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19222, 292661, 5739 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunofluorescence, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P43119, P43252, P43253 | |
Ptgir | |
Peptide (C)TQAIAPDSREMGD, corresponding to amino acid residues 290-302 of rat Prostanoid Prostacyclin receptor | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |