missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH1R (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577689
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The protein encoded by this gene is a member of the G-protein coupled receptor family 2. This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this receptor are known to be the cause of JanseNULLs metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), as well as enchodromatosis.Specifications
PTH1R (extracellular) | |
Polyclonal | |
Unconjugated | |
PTH1R | |
PTH/PTHrP type I receptor, PTH1 receptor, PTHR, PTHR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19228, 56813, 5745 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P25961, P41593, Q03431 | |
Pth1r | |
Peptide (C)DAVLYSGFTLDEAER, corresponding to amino acid residues 241-255 of rat Parathyroid Hormone 1 Receptor | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |