missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP beta/zeta/PTPRZ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80916
This item is not returnable.
View return policy
Description
PTP beta/zeta/PTPRZ Polyclonal specifically detects PTP beta/zeta/PTPRZ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PTP beta/zeta/PTPRZ | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P23471, P23471, P23471, P23471 | |
PTPRZ1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LFRHLHTVSQILPQVTSATESDKVPLHASLPVAGGDLLLEPSLAQYSDVLSTTHAASETLEFGSESGVLYKTLMFSQVEPPSSDAMMHARSSGPEPSYALSDNEGSQHIFTVSYSSAIPVHDSVGVTYQGSLFSGPSHIPIPKSSLIT | |
0.1 mL | |
Stem Cell Markers | |
5803 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3F8 CSPG phosphacan, protein tyrosine phosphatase, receptor-type, Z polypeptide 1, PTPZ, receptor-type, zeta polypeptide 1, RPTPbeta | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only