Learn More
PTRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18892-1-AP-20UL
 View more versions of this product
Product Code. 16858724
  • £135.00 / 20µL
Explore more special offers
Description
Description
The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3′ end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
Specifications
Specifications
PTRF
Polyclonal
Unconjugated
CAVIN1
Cavin 1, FKSG13, PTRF
Rabbit
Antigen Affinity Chromatography
RUO
19285, 284119, 287710
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54724, P85125, Q6NZI2
CAVIN1
PTRF Fusion Protein Ag13486
20 μL
Primary
Rat, Human, Mouse