missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PU.1 Monoclonal Antibody (E.388.3), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA515064
288.68 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
It is not recommended to aliquot this antibody.
This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
PU.1 | |
Monoclonal | |
150 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P17433, P17947 | |
SPI1 | |
Synthetic peptide corresponding to the sequence of human PU.1 protein | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
E.388.3 | |
Unconjugated | |
SPI1 | |
31 kDa transforming protein; 31 kDa-transforming protein; 31kDa Transforming Protein; Dis1; Dis-1; hCG_25181; hematopoietic transcription factor PU.1; OF; Pu.1; SFFV proviral integration 1; SFFV proviral integration 1 protein; Sfpi1; Sfpi-1; Spi1; SPI-1; Spi-1 proto-oncogene; SPIA; SPI-A; spleen focus forming virus (SFFV) proviral integration oncogene; spleen focus forming virus (SFFV) proviral integration oncogene spi1; spleen focus forming virus proviral integration oncogene spi1; Tcfpu1; Tfpu.1; Transcription factor PU.1; Transcription Factor spi1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20375, 6688 | |
-20°C | |
Liquid |
Special Offers
-
Multiple Manufacturers
New Offer Save Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PU.1 Monoclonal Antibody (E.388.3), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction