missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ PU First Grey Gloves

Catalog Number 12645293
Click to view available options
Size:
6
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Grey polyamide knitted gloves with polyurethane-coated palm and fingertips provide dexterity, a snug fit, and good grip for precision work. Honeywell Safety Products™ PU First Grey Gloves are knitted for breathability and comfort and offer excellent abrasion and tear resistance.

  • Grey, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products or dirty objects in dry, slightly greasy or dirty environments.
  • Excellent abrasion and tear resistance.
  • Silicone-free and suitable for painting environments.
  • Grey polyurethane coating on palm and fingertips for good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Ventilated back optimizes breathability.
  • Elastic wrist.
  • Grey color enhances soil proof qualities, ensuring longer life.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY II Minor risks, EN 420-2003 Basic standard, EN 388-2003 Mechanical risk, EC Category PPE 2, EC Certificate Number 0075/797/162/11/06/0281, EC Attestation Number 0075/797/162/11/06/0281
Classification Dexterity: 5
Coated Area Palm, Fingertips
Construction Seamless Knit
Description Gauge 15
Coating Material Polyurethane
Color Gray
Cut Resistance Rating 1
Material Polyamide
Product Type Gloves
Size 10
For Use With (Application) Service, Telecom, Transportation and Utility industries
Includes Elasticized wrist
Puncture Rating 1
Quantity 10Pair/PK
Tear Rating 3
Length (Metric) 25 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'