missing translation for 'onlineSavingsMsg'
Learn More

Pumice Stone, Granular, 4 to 6mm, Honeywell Fluka™

Product Code. p-7120734 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15675730

Brand: Honeywell-Fluka 51753250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Granular, 4-6 mm

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332-09-8
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
PubChem CID 76973482
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
SMILES O=[Si]=O.O=[Al]O[Al]=O

Specifications

Packaging Poly Bottle
Odor Odorless
Color Gray
Quantity 250 g
UN Number NONH for all modes of transport
Chemical Name or Material Pumice stone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.