missing translation for 'onlineSavingsMsg'
Learn More

Pumice stone, granular (approx. 1-5 mm)

Product Code. 10370134 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 10370134

Brand: Thermo Scientific Acros 390040010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1332-09-8 | C28H37FO7 | 509.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332-09-8
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
PubChem CID 76973482
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
SMILES O=[Si]=O.O=[Al]O[Al]=O

Specifications

Physical Form Granular Stones
Packaging Plastic Bottle
Color Brown/Gray
Quantity 1 kg
Chemical Name or Material Pumice stone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.