missing translation for 'onlineSavingsMsg'
Learn More
Pumice Stone, Technical, Powder, 240 Mesh, Fisher Chemical Product Code.: 10294580

Pumice Stone, Technical, Powder, 240 Mesh, Fisher Chemical

Product Code. 10294580
3 kg
Click to view available options
Quantity:
3 kg
This item is not returnable. View return policy

Product Code. 10294580

Brand: Fisher Chemical P/7760/63

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Physical Form Solid
Packaging Plastic Powder Jar
Color Gray
Quantity 3 kg
CAS 1332-09-8
Molecular Formula C28H37FO7
MDL Number 148147
InChI Key CIWBQSYVNNPZIQ-YBDIZOPJSA-N
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC
IUPAC Name [2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate
Molecular Weight (g/mol) 509.626
PubChem CID 76973482
Show More Show Less
Product Identifier
  • Pumice stone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pumice Stone, Technical, Powder, 240 Mesh, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.