Learn More
Manutan Pump, heavy duty, plastic
Product Code. 12853187
Encompass_Preferred
Product Code. 12853187
Brand: Manutan A350108
Only null left
Add to basket
Add to basket
Self-priming pistonless pump with telescopic uptake tube for transferring various liquids but not petrol or solvent. Polypropylene with LDPE parts and Viton U gland seal for 25-210ltr drums and containers. Dispensing height from drum: 229mm. 250ml delivery per stroke with discharge on down stroke.