missing translation for 'onlineSavingsMsg'
Learn More

BM Polyco™ Pura™ PVC Flock-Lined Gloves

Product Code. 12960775
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

BM Polyco™ Pura™ PVC Flock-Lined Gloves are medium-weight gloves ideal for general cleaning work or food handling

  • PVC gloves provide an excellent alternative for those allergic to natural latex proteins or sensitive to the chemical accelerators used in other types of rubber gloves
  • Anatomically shaped for additional comfort
  • Slip resistant pattern on palm enhances grip
  • Soft flock lining absorbs perspiration keeping hands cool and comfortable
  • Tested in accordance with EN1186, making them suitable for food contact
  • Also ideal for general cleaning and janitorial duties
  • Available in a wide range of colours for easy job application segregation

Specifications

Certifications/Compliance CE, EN1186, EN388, EN374-2, EN374-3
Color Blue
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material PVC
Powdered Powder Free
Size 10
Sterility Non-sterile
For Use With (Application) General Purpose Cleaning Glove
Glove Cuff Style Plain
Liner Material Flock
Lining Flock Lined
Quantity 12/Pk.
Length (Metric) 310 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'