missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex Pure-Fit Safety Glasses
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Uncoated
Description
- Reduced to the max - pure, ultralight safety spectacle with innovative side arm concept weighing only 19 grams
- Frameless lens design for a wide field of vision
- X-tended sideshield offers additional protection from hazards
- Flexible side arms deliver the perfect secure, pressure-free fit
- Nose bridge designed for all-day comfort
- Robust polycarbonate lens
- UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
- Metal-free
- Frame transparent / W 166 FT CE
- Lens PC clear, UV400 / 2C-1,2 W1 FT CE
Specifications
Specifications
Lens Coating | Uncoated |
Lens Tint | Clear |
Product Type | Ergonomic Safety Glasses |
Frame Color | Transparent |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | UV400 |
Frame Type | Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction