Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 32 to 62 in. wide
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
£2414.21 - £4973.53
Specifications
Product Type | Autoclave Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | COC |
Closure Type | Safe Seal |
Color | Clear, White |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30042194
|
Keystone Cleanroom Products
6STKRP5054SS |
50 x 54 |
£2593.06
Pack of 100 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042221
|
Keystone Cleanroom Products
6STKRP4864SS |
48 x 64 |
£2593.06
Pack of 100 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042273
|
Keystone Cleanroom Products
6STKRP4850SS |
£2820.51
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042232
|
Keystone Cleanroom Products
6STKRP4848SS |
48 x 48 |
£2771.54
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042224
|
Keystone Cleanroom Products
6STKRP4450SS |
44 x 50 |
£2820.51
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042198
|
Keystone Cleanroom Products
6STKRP4416SS |
44 x 16 |
£3322.94
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042289
|
Keystone Cleanroom Products
6STKRP4244SS |
42 x 44 |
£2664.52
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042319
|
Keystone Cleanroom Products
6STKRP4242SS |
42 x 42 |
£2602.85
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042287
|
Keystone Cleanroom Products
6STKRP4072SS |
40 x 72 |
£4387.66
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042292
|
Keystone Cleanroom Products
6STKRP4060SS |
40 x 60 |
£3134.30
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042241
|
Keystone Cleanroom Products
6STKRP4054SS |
40 x 54 |
£3010.96
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042290
|
Keystone Cleanroom Products
6STKRP4048SS |
40 x 48 |
£2900.32
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042258
|
Keystone Cleanroom Products
6STKRP4040SS |
40 x 40 |
£2588.34
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042274
|
Keystone Cleanroom Products
6STKRP4024SS |
40 x 24 |
£2588.34
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042311
|
Keystone Cleanroom Products
6STKRP3838SS |
38 x 38 |
£2508.53
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042261
|
Keystone Cleanroom Products
6STKRP3672SS |
36 x 72 |
£3134.30
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042211
|
Keystone Cleanroom Products
6STKRP3664SS |
36 x 64 |
£2820.51
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042296
|
Keystone Cleanroom Products
6STKRP3660SS |
36 x 60 |
£2806.00
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042222
|
Keystone Cleanroom Products
6STKRP3648SS |
36 x 48 |
£2633.69
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042249
|
Keystone Cleanroom Products
6STKRP3646SS |
36 x 46 |
£2508.53
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042245
|
Keystone Cleanroom Products
6STKRP3642SS |
36 x 42 |
£2414.21
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042303
|
Keystone Cleanroom Products
6STKRP3636SS |
36 x 36 |
£4726.85
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042285
|
Keystone Cleanroom Products
6STKRP3628SS |
36 x 28 |
£4385.85
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042197
|
Keystone Cleanroom Products
6STKRP3624SS |
36 x 24 |
£4385.85
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042262
|
Keystone Cleanroom Products
6STKRP3456SS |
34 x 56 |
£2820.51
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042331
|
Keystone Cleanroom Products
6STKRP3434SS |
34 x 34 |
£4770.38
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042325
|
Keystone Cleanroom Products
6STKRP3412SS |
34 x 12 |
£2601.04
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042217
|
Keystone Cleanroom Products
6STKRP3248SS |
32 x 48 |
£2695.36
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042286
|
Keystone Cleanroom Products
6STKRP3238SS |
32 x 38 |
£4973.53
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042229
|
Keystone Cleanroom Products
6STKRP3232SS |
32 x 32 |
£4509.19
Pack of 250 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30042202
|
Keystone Cleanroom Products
6STKRP6250SS |
62 x 50 |
£3604.09
Pack of 125 |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
- Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
Specifications
Autoclave Bag | |
COC | |
Clear, White | |
Steam Sterilization of Pharmaceutical Equipment | |
Non-sterile | |
Yes | |
5.3 mil |
Autoclavable | |
Safe Seal | |
Single-use | |
Tyvek | |
5.3 mil | |
121 to 125 C |