Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, up to 9 in. wide
GREENER_CHOICE
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
£1570.00 - £3840.00
Specifications
Product Type Autoclave Bag
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
View More Specs
Products 25
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30042324
View Documents
Keystone Cleanroom Products
6STKRP0205SS
2 x 5
£1570.00
Pack of 250
30042209
View Documents
Keystone Cleanroom Products
6STKRP0404SS
4 x 4
£1905.00
Pack of 250
30042226
View Documents
Keystone Cleanroom Products
6STKRP0406SS
4 x 6
£2710.00
Pack of 500
30042256
View Documents
Keystone Cleanroom Products
6STKRP0409SS
4 x 9
£3555.00
Pack of 500
30042185
View Documents
Keystone Cleanroom Products
6STKRP0410SS
4 x 10
£3590.00
Pack of 500
30042183
View Documents
Keystone Cleanroom Products
6STKRP0416SS
4 x 16
£2090.00
Pack of 250
30042301
View Documents
Keystone Cleanroom Products
6STKRP0426SS
4 x 26
£2190.00
Pack of 250
30042313
View Documents
Keystone Cleanroom Products
6STKRP0428SS
4 x 28
£2260.00
Pack of 250
30042250
View Documents
Keystone Cleanroom Products
6STKRP0430SS
4 x 30
£2385.00
Pack of 250
30042186
View Documents
Keystone Cleanroom Products
6STKRP0608SS
6 x 8
£1640.00
Pack of 250
30042240
View Documents
Keystone Cleanroom Products
6STKRP0610SS
6 x 10
£3770.00
Pack of 500
30042239
View Documents
Keystone Cleanroom Products
6STKRP0612SS
6 x 12
£3840.00
Pack of 500
30042192
View Documents
Keystone Cleanroom Products
6STKRP0615SS
6 x 15
£1990.00
Pack of 250
30042253
View Documents
Keystone Cleanroom Products
6STKRP0616SS
6 x 16
£2105.00
Pack of 250
30042305
View Documents
Keystone Cleanroom Products
6STKRP0638SS
6 x 38
£3185.00
Pack of 250
30042328
View Documents
Keystone Cleanroom Products
6STKRP0707SS
7 x 7
£2120.00
Pack of 250
30042294
View Documents
Keystone Cleanroom Products
6STKRP0709SS
7 x 9
£2175.00
Pack of 250
30042247
View Documents
Keystone Cleanroom Products
6STKRP0713SS
7 x 13
£2225.00
Pack of 250
30042326
View Documents
Keystone Cleanroom Products
6STKRP0811SS
8 x 11
£2385.00
Pack of 250
30042267
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP0815SS
8 x 15
£2485.00
Pack of 250
30042196
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP0816SS
8 x 16
£2580.00
Pack of 250
30042320
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP0820SS
8 x 20
£2560.00
Pack of 250
30042199
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP0838SS
8 x 38
£2845.00
Pack of 250
30042276
GREENER_CHOICE
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP0843SS
8 x 43
£3135.00
Pack of 250
30042279
GREENER_CHOICE
missing translation for 'viewDocuments'
Keystone Cleanroom Products
6STKRP09-512SS
9.5 x 12
£2240.00
Pack of 250
Description
Description
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Spécification
Spécification
Autoclave Bag
COC
Clear, White
Steam Sterilization of Pharmaceutical Equipment
Non-sterile
Yes
5.3 mil
Autoclavable
Safe Seal
Single-use
Tyvek
5.3 mil
121 to 125 C
Vidéos
FDS
Documents
Documents
Certificats