Learn More
Invitrogen™ PureLink™ Fast Low-Endotoxin Midi Plasmid Purification Kit
The PureLink Fast Low-Endotoxin Midi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (<1 EU/μg) plasmid DNA in about 30 minutes.
Brand: Invitrogen™ A35892
204.68 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 2.86670kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
The simple precipitation-free protocol yields up to 0.4 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.
- Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
- Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
- High yields—isolate up to 0.4 mg of high-quality plasmid DNA
- Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
Specifications
• Resuspension buffer, store at 4°C upon reciept • Lysis buffer, store at room temperature • Precipitation buffer, store at room temperature • Binding buffer, store at room temperature • Wash buffer 1, store at room temperature • Wash buffer 2 (concentrate), store at room temperature • Elution buffer, store at room temperature • Column assemblies, store at room temperature • Syringe filters, store at room temperature • Syringe plungers, store at room temperature • Collection tubes, store at room temperature |
|
Silica Column | |
Cloning, PCR | |
Midi scale input culture of 25-100 mL to obtain yields of up to 200 μg | |
Plasmid Purification Kit | |
Room Temperature |
Kit | |
Midi | |
25 Preps | |
PureLink™ | |
25 Preps | |
30 min. |
For Research Use Only. Not for use in diagnostic procedures.