Learn More
Invitrogen™ PureLink™ HiPure Expi Plasmid Megaprep Kit
The PureLink™ HiPure Plasmid Megaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.
Brand: Invitrogen™ K210008XP
140.62 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 1310-73-2 Weight : 0.40000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Description
The PureLink™ HiPure Plasmid Megaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Megaprep Kit typically isolates 4 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 500 mL of bacterial culture.
Advantages of the PureLink™ HiPure Expi Plasmid Megaprep Purification Kit include:
- High yield—isolate over 5mg of high quality plasmid DNA from a single purification using 1L of bacterial culture volume
- Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
- Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation takes as little as 90 minutes
HiPure Expi anion exchange chromatography for plasmid purification
PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
• Equilibration Buffer • Resuspension Buffer • Lysis Buffer • Precipitation Buffer • RNase A • Wash Buffer • Elution Buffer • TE Buffer • Expi HiPure Mega Filtration Column • Expi HiPure Mega DNA Binding Column Store all components at room temperature upon receipt. |
|
Anion Exchange Resin | |
Manual Protocols | |
4 mg plasmid DNA | |
Plasmid MegaPrep Kit | |
Approved for shipment at Room Temperature or on Wet Ice | |
2 hr. |
Kit | |
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription | |
4 Preps | |
PureLink™ | |
4 Preps | |
Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.