missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HALYARD™ PUREZERO™ HG3 Light Blue Nitrile Gloves



Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination
- Nickel-free
- Powder-free
- Silicone-free
- Latex-free and no phtalates
- High cleanliness
- Low endotoxin levels
- Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
Specifications
Specifications
Certifications/Compliance | ISO 3, ISO 4, ISO 5, ISO 6, ISO 7 |
Certifications/Compliance (Europe) | GMP Grade A/B/C/D |
Chemotherapeutic Rated | ASTM D6978 |
Chlorinated | Yes |
Cleanroom Class | Class 10 (ISO 4) |
Accelerator Free | Yes |
AQL | 1 |
Color | Light Blue |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction