missing translation for 'onlineSavingsMsg'
Learn More
HALYARD™ PUREZERO™ HG3 White Sterile Nitrile Gloves Product Code.: 17374330
In Stock
to see stock.
Add to basket

HALYARD™ PUREZERO™ HG3 White Sterile Nitrile Gloves
GREENER_CHOICEBIOPHARMA

Product Code. 17374330
6
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
10

Product Code. 17374330

Brand: HALYARD™ CLN323260

In Stock
to see stock.
Add to basket
Add to basket

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.

  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no Phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
  • Backed up by a vertically integrated supply chain
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
    • Renewable Energy
    • Source Reduction

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 10 (ISO 4)
Accelerator Free Yes
AQL 1
Color White
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.16 mm
Hand Specific
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 6
Sterility Sterile
Thickness (English) 6 mil
Thickness (Metric) 0.16 mm
Manufacturing/QMS Audit Standards ISO 9001:2015, EN13485:2016
Model CLN323260
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSM420325X Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberCLN323260
Lot NumberSM421825X Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberCLN323260
Lot NumberSM215525X Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberCLN323260
Lot NumberSM221725X Certificate TypeCertificate of Irradiation Date11/01/2023 Catalog NumberCLN323260
Lot NumberSM215325X Certificate TypeCertificate of Irradiation Date19/12/2022 Catalog NumberCLN323260
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HALYARD™ PUREZERO™ HG3 White Sterile Nitrile Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.