missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purified Water, Reagecon
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
5 L
Packaging:
HDPE container
Description
- Meets water reference standard, Water Reference Material
- Packaged in a 5L HDPE container
Specifications
pH | 7 |
CAS | 7732-18-5 |
Color | Clear, Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
CAS Max % | 1 |
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
Grade | Purified |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'