missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purine, 99%, Thermo Scientific Chemicals
£138.00 - £469.00
Chemical Identifiers
CAS | 120-73-0 |
---|---|
MDL Number | MFCD00079221 |
InChI Key | KDCGOANMDULRCW-UHFFFAOYSA-N |
Synonym | purine, 9h-purine, 1h-purine, isopurine, beta-purine, 7h-imidazo 4,5-d pyrimidine, imidazo 4,5-d pyrimidine, 3,5,7-triazaindole, 9h-purine van, 3h-purine |
PubChem CID | 1044 |
ChEBI | CHEBI:17258 |
IUPAC Name | 7H-purine |
SMILES | C1=C2C(=NC=N1)N=CN2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10666142
|
Thermo Scientific Acros
131670010 |
1 g | Glass bottle |
£138.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360904
|
Thermo Scientific Acros
131670050 |
5 g | Glass bottle |
£469.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-73-0 | |
KDCGOANMDULRCW-UHFFFAOYSA-N | |
1044 | |
7H-purine |
MFCD00079221 | |
purine, 9h-purine, 1h-purine, isopurine, beta-purine, 7h-imidazo 4,5-d pyrimidine, imidazo 4,5-d pyrimidine, 3,5,7-triazaindole, 9h-purine van, 3h-purine | |
CHEBI:17258 | |
C1=C2C(=NC=N1)N=CN2 |
Specifications
120-73-0 | |
White to Beige | |
98.5% min. (HPLC) | |
MFCD00079221 | |
15, 8052 | |
Solubility in water: 400g/L water (20°C). Other solubilities: soluble in alcohol,toluene,acetone,hot ethylacet | |
C1=C2C(=NC=N1)N=CN2 | |
120.11 | |
CHEBI:17258 | |
99% | |
Purine |
214°C to 217°C | |
Authentic | |
C5H4N4 | |
26, 354 | |
purine, 9h-purine, 1h-purine, isopurine, beta-purine, 7h-imidazo 4,5-d pyrimidine, imidazo 4,5-d pyrimidine, 3,5,7-triazaindole, 9h-purine van, 3h-purine | |
KDCGOANMDULRCW-UHFFFAOYSA-N | |
7H-purine | |
1044 | |
120.11 | |
Crystalline Powder or Crystals |
Safety and Handling
missing translation for 'einecsNumber' : 204-421-2
RUO – Research Use Only