Learn More
Corning™ Puromycin Dihydrochloride
Packaged in plastic bottles
Brand: Corning™ 61-385-RA
Code : NEW
Additional Details : CAS Number : 64-75-5 Weight : 0.01000kg
Specifications
-5° to -25°C | |
Antibiotic | |
100 mg |
Powder | |
Plastic Bottle |
Packaged in plastic bottles
Brand: Corning™ 61-385-RA
Code : NEW
Additional Details : CAS Number : 64-75-5 Weight : 0.01000kg
-5° to -25°C | |
Antibiotic | |
100 mg |
Powder | |
Plastic Bottle |