Learn More
MP Biomedicals™ Purple Screw Caps
Use with empty FastPrep™ tubes. Purple Caps are screw caps with an O-ring for use with 2 mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix E tubes for prepare samples for homogenization.
Brand: MP Biomedicals™ 115064010
Code : NEW
Additional Details : Weight : 1.70000kg
Description
- Purple Caps fit empty FastPrep™ tubes and are used in FastPrep™ systems.
- Add sample to empty tube, add Lysing Matrix E material and lysing buffer, and apply cap. Process using FastPrep™ instrument or other bead-beater to lyse, grind or homogenize sample.
- Purple caps are also found in the Fast DNA™ Spin Kit for Soil and Fast RNA™ Pro Soil Kit.
Specifications
Purple | |
Store at Room Temperature (15°C to 30°C) | |
Screw Cap |
2 mL | |
5000 Pk. |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.