Learn More
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
BIOPHARMA
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
Marca:  PURUS™ PM 3660 34 B-HT
 Ver más versiones de este producto
Código de producto. 15312002
  • £155.00 / Pack of 4
Estimated Shipment: 03-04-2025
to see stock.
Explore más ofertas especiales
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
  • Earth Friendly containing GMO free, renewable plant based Cardia Biohybrid™ material that reduces CO2 emissions
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Certifications
Manufactured in ISO 9001 certified facility
Specifications
Specifications
Manufactured in an ISO 9001 certified facility
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
High Tack
Blue
914 x 1524 mm
60 Sheets/Mat, 4 Mats/Cs.
30 layer
LDPE Blend
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats > 36 x 60 in.; 30 layer; Blue