Learn More
PURUS™ Frames for EnviroTack™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
Brand:  PURUS™ PF 2038 01 W 00
72.88 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15342002
  • £86.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS™ EnviroTack™ mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications
Specifications
508 x 965 mm
457 x 914 mm PURUS™ Biohybrid™ Mats
White
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
3.175 mm
Polystyrene