Learn More
RL Enterprises™ Cork Borer Push Rod
Push rod for clearing borers.
Brand: RL Enterprises™ 3400/ROD
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Manufactured from brass and nickel-plated
- Sizes numbered consecutively from 1 upwards in each set
Specifications
Push Rod only | |
Nickel-plated brass | |
Cork | |
Each |