missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ PVC Bulk Wrapped Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
£89.65
Specifications
Color | Clear |
---|---|
Material | PVC |
Sterility | Non-sterile |
Type | Reagent Reservoir |
For Use With (Application) | Reagent Pipetting |
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Quality certificate available on request
- Disposable
Specifications
Clear | |
Non-sterile | |
Reagent Pipetting | |
100 Pk. | |
1 | |
Convoluted |
PVC | |
Reagent Reservoir | |
Bulk Wrapped | |
Not Autoclavable | |
Single-use |