Learn More
Gilson™ PVC Tubing for MF1 and MF4 pump head
For use with MF1 and MF4 pump head
£85.80 - £89.40
Specifications
Autoclavable Autoclavable
Material PVC
Overall Length (Metric) 300 cm
For Use With For MF1 and MF4 pump heads
For Use With (Application) general use
View More Specs
Products 7
Product Code Brand Diameter (Metric) Inner Flow Rate Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Flow Rate Price Quantity & Availability  
10464712
View Documents Promotion Details
Gilson™
F117952
0.25 mm 0.574mL/min.
£85.80
3m
In Stock
to see stock.
Add to basket
10011972
View Documents
Gilson™
F117953
0.38 mm 1.22mL/min.
£89.40
3m
In Stock
to see stock.
Add to basket
10061992
View Documents Promotion Details
Gilson™
F117965
2.06 mm 25mL/min.
£85.80
3m
In Stock
to see stock.
Add to basket
10021982
View Documents Promotion Details
Gilson™
F117958
1.02 mm 6mL/min.
£85.80
3m
In Stock
to see stock.
Add to basket
10541103
View Documents Promotion Details
Gilson™
F117962
1.52 mm 14mL/min.
£89.40
3m
In Stock
to see stock.
Add to basket
10327232
View Documents
Gilson™
F117968
2.79 mm 39mL/min.
£85.80
3m
In Stock
to see stock.
Add to basket
10674923
View Documents Promotion Details
Gilson™
F117969
3.18 mm 47mL/min.
£88.80
3m
In Stock
to see stock.
Add to basket
Description
Description
  • PVC
Specifications
Specifications
Autoclavable
300 cm
general use
Low
94°C
PVC
For MF1 and MF4 pump heads
1 Each
Transparent
Extension Pump Tubing
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Gilson™ PVC Tubing for MF1 and MF4 pump head