missing translation for 'onlineSavingsMsg'
Learn More

Dostmann Electronic™ PVC Precision Probes

Rich, robust accessory for hand-held meter Series P700 that is ideal for use in many laboratory, scientific, and research environments.

£216.00 - £307.00

Specifications

Type Probe, Precision
Material PVC
Length (Metric) 300 mm
View More Specs

Products 2
Product Code Brand Length (Metric) Sensor Types Price Quantity & Availability  
Product Code Brand Length (Metric) Sensor Types Price Quantity & Availability  
10341492
View Documents
Dostmann Electronic™
6000-1019
300 mm Class 1/3
£216.00
Each
Please to purchase this item. Need a web account? Register with us today!
11854402
View Documents
Dostmann Electronic™
6000-1074
300 mm Class 1/10
£307.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  •  Accessories for hand-held meters Series P700
  • Immersion probe Pt100, -200 to +450°C, ±0.03°C, 300x3mm 
Specifications

Specifications

Probe, Precision
PVC
300 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.