Learn More
Bolle Protection™ PVC Visor Goggle
Use the Bolle Protection™ PVC Visor Goggle for all-round vision and for use to cover prescription glasses. It has an overflow chute for liquids and is compatible with a half-mask respirator.
Brand: Bolle Protection™ BLV
7.26 GBP valid until 2024-12-31
Use promo code "23287" to get your promotional price.
Code : S1
Additional Details : Weight : 0.07200kg
Description
- All-round vision
- Adjustable strap
- Overflow chute for liquids
- Fits over prescription glasses
- Compatible with half-mask respirator
Specifications
Blue | |
PVC | |
Visor accessory. adjustable strap |