missing translation for 'onlineSavingsMsg'
Learn More

Merck PVDF Durapore™ Media Membrane Filter Sheets

Brand:  Merck HVHP00010

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 10488561

  • £464.00 / Each
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Membrane Filter
0.45 μm
Ideal filter to genetic engineering products and other protein eradication because of the non-specific proteins of adsorption is very low
Membrane Filter
PVDF
HVHP
1Pack
304 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers