Learn More
Cole-Parmer™ PVDF Manifold, 8 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Brand: Cole-Parmer™ 31523-53
This item is not returnable.
View return policy
Beschreibung
- Constructed of durable PVDF.
- Ideal for applications requiring excellent chemical compatibly.
- Available in a variety of configurations
Spezifikation
Manifold | |
8.75 in. | |
-65°F | |
1/2 in. x 3/8 in. | |
3/8 in. NPT(F) | |
1.5 in. | |
1.5 in. | |
230°F |
PVDF | |
22.23 cm | |
-54°C | |
1/2 in. NPT(F) | |
3.81 cm | |
3.81 cm | |
110 psi | |
110°C |