Learn More
Karl Hecht™ Pycnometers According to Gay-Lussac
For the measurement of liquid density in the laboratory.
Brand:  Karl Hecht™ 42571008
 View more versions of this product
Product Code. 10758102
  • £61.60 / Each
Explore more special offers
Description
Description
  • With ground-in stopper
  • Precisely adjusted
  • Available in different capacities
Specifications
Specifications
Pycnometer acc. to Gay-Lussac
5 mL
Measuring liquid density
With ground-in stopper