Learn More
Pyraclostrobin, Honeywell Fluka™
Brand: Honeywell Fluka R1734-100MG
Additional Details : Weight : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Pyraclostrobin | |
≤ 100% | |
387.82 g/mol | |
200°C (Decomposition) |
175013-18-0 | |
Methyl {2-[1-(4-chlorophenyl)-1H-pyrazol-3-yloxymethyl]phenyl}methoxycarbamate | |
64°C | |
100 mg |