missing translation for 'onlineSavingsMsg'
Learn More
Pyrazine-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11459617

Pyrazine-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11459617
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11459617

Brand: Thermo Scientific Alfa Aesar B22108.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-01-0
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00006131
InChI Key ZUCRGHABDDWQPY-UHFFFAOYSA-N
Synonym 2,3-pyrazinedicarboxylic acid, pyrazine-2,3-dicarboxylate, 2,3-dicarboxypyrazine, 2,3-pyrazine dicarboxylic acid, unii-qnn35wof29, 2,3-pyrazinedicarboxylicacid, qnn35wof29, 2,3-pyrazinedcarboxylicacid, pubchem8610, acmc-209qye
PubChem CID 66628
IUPAC Name pyrazine-2,3-dicarboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)C(=O)O

Specifications

Melting Point ∼188°C (decomposition)
Quantity 5 g
Beilstein 147982
Merck Index 14,7958
Formula Weight 168.11
Percent Purity 98%
Chemical Name or Material Pyrazine-2,3-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazine-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.