missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 50°C to 56°C |
Color | White |
Density | 1.031 g/mL |
Boiling Point | 113°C to 116°C |
Flash Point | 55°C (131°F) |
Odor | Strong |
Refractive Index | 1.5235 |
Quantity | 50 g |
UN Number | UN1325 |
Beilstein | 103905 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'