Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Manufactured from borosilicate glass
Brand:  Pyrex 1541/12D
 View more versions of this product
Product Code. 11337884
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Compliance
ISO 4796
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2000 mL
Borosilicate Glass
Aspirator Bottle
28 mm
248 mm
138 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products