missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Aspirator Bottle with Ground Glass Side Socket

Manufactured from borosilicate glass

Brand:  Pyrex 1541/18D

282.50 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11357884

  • £304.00 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from borosilicate glass

  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers

Compliance

ISO 4796

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10,000 mL
Borosilicate Glass
Aspirator Bottle
59 mm
398 mm
227 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.